The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015693.3 | NP_056508.2 | 9 - 10 | 1610 | 64 |
XM_011531844.2 | XP_011530146.1 | 9 - 10 | 1634 | 64 | |
XM_011531846.1 | XP_011530148.1 | 9 - 10 | 1582 | 64 | |
XM_011531847.1 | XP_011530149.1 | 9 - 10 | 1019 | 64 | |
XM_011531848.2 | XP_011530150.1 | 4 - 5 | 445 | 64 | |
XM_011531849.2 | XP_011530151.1 | 9 - 10 | 1637 | 64 | |
XM_017008025.1 | XP_016863514.1 | 6 - 7 | 553 | 64 | |
GenBank mRNA | AB033110.1 | - | 9 - 10 | 1577 | 64 |
AK123320.1 | - | 2 - 3 | 950 | 64 | |
AK291481.1 | - | 9 - 10 | 1592 | 64 | |
BC051698.1 | - | 11 - 12 | 4078 | 64 | |
BC130611.1 | - | 9 - 10 | 1576 | 64 |