The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303270.1 | NP_001290199.1 | 3 - 4 | 317 | 71 |
NM_020877.3 | NP_065928.2 | 3 - 4 | 1691 | 71 | |
XM_011523663.1 | XP_011521965.1 | 3 - 4 | 313 | 71 | |
XM_011523664.2 | XP_011521966.1 | 4 - 5 | 462 | 71 | |
XM_011523666.1 | XP_011521968.1 | 3 - 4 | 313 | 71 | |
XM_011523667.2 | XP_011521969.1 | 4 - 5 | 462 | 71 | |
XM_011523670.2 | XP_011521972.1 | 3 - 4 | 313 | 71 | |
XM_017024219.1 | XP_016879708.1 | 3 - 4 | 313 | 71 | |
GenBank mRNA | AB040936.2 | - | 3 - 4 | 1691 | 71 |
AK292607.1 | - | 3 - 4 | 313 | 71 | |
BC034225.1 | - | 3 - 4 | 316 | 71 |