The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080414.3 | NP_001073883.2 | 15 - 16 | 2891 | 100 |
XM_005267691.4 | XP_005267748.1 | 15 - 16 | 3545 | 100 | |
XM_011536796.2 | XP_011535098.1 | 15 - 16 | 2845 | 100 | |
XM_017021335.1 | XP_016876824.1 | 15 - 16 | 3545 | 100 |