Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDHR1
Entrez Gene ID
92211
Gene Name
cadherin related family member 1
Gene Aliases
CORD15, PCDH21, PRCAD, RP65
Chromosome Location
Chr.10: 84194635 - 84219621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171971.2 NP_001165442.1 12 - 13 1445 93
NM_033100.3 NP_149091.1 12 - 13 1445 93
XM_011540337.1 XP_011538639.1 11 - 12 1936 93
XM_011540338.1 XP_011538640.1 11 - 12 1936 93
XM_011540339.1 XP_011538641.1 12 - 13 1251 93
XM_011540340.2 XP_011538642.1 11 - 12 1936 93
GenBank mRNA AB053448.1 - 12 - 13 1318 93
AB593154.1 - 12 - 13 1445 93
AB593155.1 - 12 - 13 1431 93
BC038799.1 - 12 - 13 1435 93

View Full Product Details