The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099433.1 | NP_001092903.1 | 3 - 4 | 1111 | 56 |
NM_001306133.1 | NP_001293062.1 | 3 - 4 | 901 | 56 | |
NM_144720.3 | NP_653321.1 | 3 - 4 | 1111 | 56 | |
XM_017007788.1 | XP_016863277.1 | 3 - 4 | 1075 | 56 | |
XM_017007789.1 | XP_016863278.1 | 3 - 4 | 899 | 56 | |
XM_017007790.1 | XP_016863279.1 | 3 - 4 | 1051 | 56 | |
XM_017007791.1 | XP_016863280.1 | 3 - 4 | 1075 | 56 | |
XM_017007792.1 | XP_016863281.1 | 3 - 4 | 1075 | 56 | |
XM_017007793.1 | XP_016863282.1 | 3 - 4 | 1075 | 56 | |
XM_017007794.1 | XP_016863283.1 | 3 - 4 | 1075 | 56 | |
XM_017007795.1 | XP_016863284.1 | 3 - 4 | 1075 | 56 | |
XM_017007796.1 | XP_016863285.1 | 3 - 4 | 1075 | 56 | |
GenBank mRNA | AK056126.1 | - | 3 - 4 | 1075 | 56 |
AK124718.1 | - | 3 - 4 | 899 | 56 | |
AK294356.1 | - | 4 - 5 | 728 | 56 | |
AK307983.1 | - | 2 - 3 | 326 | 56 | |
AM412307.1 | - | 3 - 4 | 1075 | 56 | |
AM412308.1 | - | 3 - 4 | 1075 | 56 | |
AM412309.1 | - | 3 - 4 | 1075 | 56 | |
AY382340.1 | - | 2 - 3 | 625 | 56 | |
BC047075.1 | - | 3 - 4 | 831 | 56 |