Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA18
Entrez Gene ID
132671
Gene Name
spermatogenesis associated 18
Gene Aliases
Mieap, SPETEX1
Chromosome Location
Chr.4: 52051331 - 52097305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297608.1 NP_001284537.1 8 - 9 1637 72
NM_145263.3 NP_660306.1 9 - 10 1733 72
NR_123728.1 9 - 10 1733 72
XM_005265716.1 XP_005265773.1 7 - 8 1439 72
XM_005265718.1 XP_005265775.1 5 - 6 1221 72
XM_011534369.1 XP_011532671.1 8 - 9 1535 72
XM_011534370.1 XP_011532672.1 9 - 10 1659 72
XM_011534372.1 XP_011532674.1 6 - 7 1313 72
XM_017007731.1 XP_016863220.1 7 - 8 1131 72
GenBank mRNA AB465501.1 - 9 - 10 1359 72
AB465502.1 - 8 - 9 1263 72
AK057468.1 - 9 - 10 1645 72
AK098625.1 - 6 - 7 958 72
AK304386.1 - 8 - 9 1552 72
AK314799.1 - 9 - 10 1603 72
BC025396.1 - 9 - 10 1640 72
BC037886.1 - 3 - 4 412 72

View Full Product Details