The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000565.3 | NP_000556.1 | 3 - 4 | 900 | 65 |
NM_001206866.1 | NP_001193795.1 | 3 - 4 | 900 | 65 | |
NM_181359.2 | NP_852004.1 | 3 - 4 | 900 | 65 | |
XM_005245139.1 | XP_005245196.1 | 3 - 4 | 859 | 65 | |
XM_005245140.2 | XP_005245197.1 | 3 - 4 | 859 | 65 | |
XM_006711298.1 | XP_006711361.1 | 3 - 4 | 859 | 65 | |
XM_006711299.3 | XP_006711362.1 | 3 - 4 | 859 | 65 | |
XM_017001199.1 | XP_016856688.1 | 3 - 4 | 859 | 65 | |
XM_017001200.1 | XP_016856689.1 | 3 - 4 | 859 | 65 | |
XM_017001201.1 | XP_016856690.1 | 3 - 4 | 859 | 65 | |
GenBank mRNA | AK223582.1 | - | 3 - 4 | 721 | 65 |
AK293013.1 | - | 3 - 4 | 514 | 65 | |
AK312730.1 | - | 3 - 4 | 511 | 65 | |
BC035327.1 | - | 3 - 4 | 475 | 65 | |
BC089410.1 | - | 3 - 4 | 870 | 65 | |
BC132684.1 | - | 3 - 4 | 713 | 65 | |
BC132686.1 | - | 3 - 4 | 713 | 65 | |
X12830.1 | - | 3 - 4 | 900 | 65 | |
X58298.1 | - | 3 - 4 | 514 | 65 |