The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184.3 | NP_001175.2 | 5 - 6 | 1465 | 81 |
XM_011512924.1 | XP_011511226.1 | 5 - 6 | 1432 | 81 | |
XM_011512926.2 | XP_011511228.1 | 5 - 6 | 1432 | 81 | |
GenBank mRNA | AK307402.1 | - | 5 - 6 | 1432 | 81 |
U49844.1 | - | 5 - 6 | 1448 | 81 | |
U76308.1 | - | 5 - 6 | 1422 | 81 | |
Y09077.1 | - | 5 - 6 | 1422 | 81 |