Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOCK1
Entrez Gene ID
1793
Gene Name
dedicator of cytokinesis 1
Gene Aliases
DOCK180, ced5
Chromosome Location
Chr.10: 126905409 - 127452517 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290223.1 NP_001277152.1 29 - 30 3154 113
NM_001380.4 NP_001371.1 29 - 30 3091 113
XM_011539422.2 XP_011537724.1 29 - 30 3132 113
XM_011539424.1 XP_011537726.1 29 - 30 3042 113
XM_017015813.1 XP_016871302.1 29 - 30 3064 113
XM_017015816.1 XP_016871305.1 12 - 13 1312 113
GenBank mRNA BC146857.1 - 29 - 30 3044 113
D50857.1 - 29 - 30 3005 113

View Full Product Details