The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111045.1 | NP_001104515.1 | 3 - 4 | 889 | 81 |
NM_001111046.1 | NP_001104516.1 | 3 - 4 | 771 | 81 | |
NM_001111047.1 | NP_001104517.1 | 3 - 4 | 768 | 81 | |
NM_003914.3 | NP_003905.1 | 3 - 4 | 892 | 81 | |
XM_011535294.2 | XP_011533596.1 | 3 - 4 | 1723 | 81 | |
XM_011535295.2 | XP_011533597.1 | 3 - 4 | 1720 | 81 | |
XM_011535296.2 | XP_011533598.1 | 3 - 4 | 548 | 81 | |
GenBank mRNA | AK301897.1 | - | 3 - 4 | 664 | 81 |
AK314261.1 | - | 3 - 4 | 892 | 81 | |
AK316392.1 | - | 3 - 4 | 691 | 81 | |
BC035479.1 | - | 3 - 4 | 638 | 81 | |
BC036346.1 | - | 3 - 4 | 638 | 81 | |
BT019577.1 | - | 3 - 4 | 533 | 81 | |
U66838.1 | - | 3 - 4 | 671 | 81 | |
U97680.1 | - | 2 - 3 | 381 | 81 |