Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTM3
Entrez Gene ID
2947
Gene Name
glutathione S-transferase mu 3
Gene Aliases
GST5, GSTB, GSTM3-3, GTM3
Chromosome Location
Chr.1: 109733932 - 109741038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000849.4 NP_000840.2 4 - 5 503 83
NR_024537.1 3 - 4 427 83
GenBank mRNA AB209816.1 - 3 - 4 212 83
AK313444.1 - 3 - 4 221 83
BC000088.2 - 3 - 4 208 83
BC008790.2 - 4 - 5 503 83
BE392471.1 - 4 - 5 484 83
BT019945.1 - 3 - 4 193 83
CR542108.1 - 3 - 4 193 83
EF363096.1 - 3 - 4 210 83
J05459.1 - 3 - 4 210 83

View Full Product Details