Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLNC
Entrez Gene ID
2318
Gene Name
filamin C
Gene Aliases
ABP-280, ABP280A, ABPA, ABPL, FLN2, MFM5, MPD4
Chromosome Location
Chr.7: 128830429 - 128859274 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127487.1 NP_001120959.1 22 - 23 4174 77
NM_001458.4 NP_001449.3 22 - 23 4174 77
GenBank mRNA AB371585.1 - 22 - 23 4194 77
AF089841.1 - 22 - 23 4014 77
AF146692.1 - 22 - 23 77
AJ012737.1 - 22 - 23 3983 77

View Full Product Details