The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003005.3 | NP_002996.2 | 16 - 17 | 2553 | 118 |
XM_005245439.1 | XP_005245496.1 | 15 - 16 | 2430 | 118 | |
GenBank mRNA | BC068533.1 | - | 16 - 17 | 2536 | 118 |
M25322.1 | - | 16 - 17 | 2526 | 118 |