The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034.3 | NP_001025.1 | 1 - 2 | 163 | 79 |
NM_001165931.1 | NP_001159403.1 | 1 - 2 | 331 | 79 | |
GenBank mRNA | AK123010.1 | - | 1 - 2 | 160 | 79 |
AK313719.1 | - | 1 - 2 | 160 | 79 | |
BC001886.1 | - | 1 - 2 | 144 | 79 | |
BC030154.1 | - | 1 - 2 | 149 | 79 | |
S40301.1 | - | 1 - 2 | 294 | 79 | |
X59618.1 | - | 1 - 2 | 294 | 79 |