The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002147.3 | NP_002138.1 | 1 - 2 | 619 | 68 |
GenBank mRNA | AK314964.1 | - | 1 - 2 | 676 | 68 |
BC117247.1 | - | 1 - 2 | 580 | 68 | |
BE621135.1 | - | 1 - 2 | 416 | 68 | |
M92299.1 | - | 1 - 2 | 835 | 68 |