Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOVA1
Entrez Gene ID
4857
Gene Name
NOVA alternative splicing regulator 1
Gene Aliases
Nova-1
Chromosome Location
Chr.14: 26445883 - 26597754 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002515.2 NP_002506.2 1 - 2 455 104
NM_006489.2 NP_006480.2 1 - 2 455 104
NM_006491.2 NP_006482.1 1 - 2 455 104
XM_017021344.1 XP_016876833.1 1 - 2 220 104
GenBank mRNA AK226114.1 - 1 - 2 172 104
AK289639.1 - 1 - 2 141 104
AK291022.1 - 1 - 2 278 104
AK291468.1 - 1 - 2 156 104
AK301526.1 - 1 - 2 137 104
BC075038.2 - 1 - 2 160 104
BC075039.2 - 1 - 2 161 104
CN409914.1 - 1 - 2 458 104
U04840.1 - 1 - 2 197 104

View Full Product Details