Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAN2C1
Entrez Gene ID
4123
Gene Name
mannosidase alpha class 2C member 1
Gene Aliases
MAN6A8, MANA, MANA1
Chromosome Location
Chr.15: 75355792 - 75368680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256494.1 NP_001243423.1 12 - 13 1509 73
NM_001256495.1 NP_001243424.1 12 - 13 1509 73
NM_001256496.1 NP_001243425.1 10 - 11 1212 73
NM_006715.3 NP_006706.2 12 - 13 1509 73
XM_005254384.2 XP_005254441.1 12 - 13 1562 73
XM_017022186.1 XP_016877675.1 11 - 12 1857 73
XM_017022187.1 XP_016877676.1 11 - 12 1857 73
XM_017022188.1 XP_016877677.1 10 - 11 1265 73
GenBank mRNA AF044414.2 - 12 - 13 1521 73
AK225145.1 - 10 - 11 1189 73
AK311065.1 - 11 - 12 1361 73
AL136876.1 - 12 - 13 1470 73
BC050550.1 - 12 - 13 1476 73
BC080191.1 - 12 - 13 1474 73

View Full Product Details