The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039592.1 | NP_001034681.1 | 1 - 2 | 159 | 87 |
NM_172312.1 | NP_758516.1 | 1 - 2 | 159 | 87 | |
GenBank mRNA | AK302313.1 | - | 1 - 2 | 67 | 87 |
BI757195.1 | - | 1 - 2 | 146 | 87 | |
S83157.1 | - | 1 - 2 | 160 | 87 | |
U12978.2 | - | 1 - 2 | 160 | 87 |