The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287758.1 | NP_001274687.1 | 6 - 7 | 680 | 66 |
NM_001287759.1 | NP_001274688.1 | 6 - 7 | 680 | 66 | |
NM_001287760.1 | NP_001274689.1 | 6 - 7 | 680 | 66 | |
NM_001847.3 | NP_001838.2 | 6 - 7 | 575 | 66 | |
NM_033641.3 | NP_378667.1 | 6 - 7 | 680 | 66 | |
XM_006724617.3 | XP_006724680.1 | 6 - 7 | 569 | 66 | |
XM_011530852.2 | XP_011529154.1 | 6 - 7 | 569 | 66 | |
XM_011530853.2 | XP_011529155.1 | 4 - 5 | 1285 | 66 | |
XM_011530854.2 | XP_011529156.1 | 6 - 7 | 570 | 66 | |
GenBank mRNA | AW082083.1 | - | 8 - 9 | 66 | |
BC000874.1 | - | 6 - 7 | 614 | 66 | |
BC140855.1 | - | 6 - 7 | 576 | 66 | |
BC144669.1 | - | 6 - 7 | 576 | 66 | |
CN313347.1 | - | 6 - 7 | 683 | 66 | |
D21337.1 | - | 6 - 7 | 680 | 66 | |
L22763.1 | - | 6 - 7 | 558 | 66 | |
U04845.1 | - | 6 - 7 | 558 | 66 |