The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301030.1 | NP_001287959.1 | 10 - 11 | 1313 | 68 |
NM_002088.4 | NP_002079.3 | 10 - 11 | 1303 | 68 | |
XM_005258821.3 | XP_005258878.1 | 11 - 12 | 2216 | 68 | |
XM_011526862.2 | XP_011525164.1 | 11 - 12 | 2216 | 68 | |
XM_011526863.2 | XP_011525165.1 | 11 - 12 | 2216 | 68 | |
XM_011526865.1 | XP_011525167.1 | 12 - 13 | 2717 | 68 | |
XM_011526867.1 | XP_011525169.1 | 13 - 14 | 2942 | 68 | |
XM_011526868.1 | XP_011525170.1 | 12 - 13 | 2695 | 68 | |
XM_011526869.2 | XP_011525171.1 | 11 - 12 | 2216 | 68 | |
XM_011526870.2 | XP_011525172.1 | 11 - 12 | 2216 | 68 | |
XM_011526871.2 | XP_011525173.1 | 11 - 12 | 2216 | 68 | |
XM_017026713.1 | XP_016882202.1 | 13 - 14 | 2942 | 68 | |
GenBank mRNA | AJ249209.1 | - | 10 - 11 | 1303 | 68 |
BC037358.1 | - | 9 - 10 | 1056 | 68 | |
S40369.1 | - | 10 - 11 | 1268 | 68 |