Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRG1
Entrez Gene ID
4733
Gene Name
developmentally regulated GTP binding protein 1
Gene Aliases
NEDD3
Chromosome Location
Chr.22: 31399553 - 31434186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004147.3 NP_004138.1 1 - 2 172 76
GenBank mRNA AF078103.1 - 1 - 2 102 76
AJ005940.1 - 1 - 2 106 76
AK315659.1 - 1 - 2 130 76
BC019285.1 - 1 - 2 111 76
BC020803.1 - 1 - 2 106 76
CR456488.1 - 1 - 2 83 76

View Full Product Details