Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAALAD2
Entrez Gene ID
10003
Gene Name
N-acetylated alpha-linked acidic dipeptidase 2
Gene Aliases
GCPIII, GPCIII, NAADALASE2, NAALADASE2
Chromosome Location
Chr.11: 90131694 - 90193577 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005467.3 NP_005458.1 9 - 10 1184 123
XM_017017043.1 XP_016872532.1 11 - 12 1350 123
XM_017017045.1 XP_016872534.1 9 - 10 1187 123
GenBank mRNA AJ012370.1 - 9 - 10 1120 123
AK075390.1 - 9 - 10 1159 123
BC027594.1 - 8 - 9 2140 123
BC096316.3 - 9 - 10 1096 123
BC096317.3 - 9 - 10 1095 123
BC096318.3 - 9 - 10 1095 123

View Full Product Details