Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DNAJB6
- Entrez Gene ID
-
10049
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B6
- Gene Aliases
-
DJ4, DnaJ, HHDJ1, HSJ-2, HSJ2, LGMD1D, LGMD1E, MRJ, MSJ-1
- Chromosome Location
-
Chr.7: 157337004 - 157417439 on Build GRCh38
- Gene Symbol
- APOL3
- Entrez Gene ID
-
80833
- Gene Name
- apolipoprotein L3
- Gene Aliases
-
APOLIII, CG121, CG12_1, apoL-III
- Chromosome Location
-
Chr.22: 36140323 - 36166177 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK297892.1 |
- |
- |
330 |
88 |
- Gene Symbol
- TMEM135
- Entrez Gene ID
-
65084
- Gene Name
- transmembrane protein 135
- Gene Aliases
-
PMP52
- Chromosome Location
-
Chr.11: 87037844 - 87328834 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF080569.1 |
- |
3 - 4 |
231 |
88 |
View Full Product Details