The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012192.3 | NP_036324.1 | 2 - 3 | 208 | 91 |
GenBank mRNA | AF150105.1 | - | 2 - 3 | 197 | 91 |
AF152355.1 | - | 2 - 3 | 138 | 91 | |
AF183415.1 | - | 2 - 3 | 196 | 91 | |
AK002130.1 | - | 2 - 3 | 169 | 91 | |
AK311763.1 | - | 2 - 3 | 154 | 91 | |
BC011014.1 | - | 2 - 3 | 150 | 91 | |
DA290694.1 | - | 2 - 3 | 208 | 91 |