The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014159.6 | NP_054878.5 | 6 - 7 | 4888 | 113 |
XM_011533631.2 | XP_011531933.1 | 7 - 8 | 6384 | 113 | |
XM_011533632.2 | XP_011531934.1 | 6 - 7 | 4960 | 113 | |
XM_011533633.2 | XP_011531935.1 | 7 - 8 | 6386 | 113 | |
XM_011533634.1 | XP_011531936.1 | 6 - 7 | 4913 | 113 | |
GenBank mRNA | AB051519.2 | - | 4 - 5 | 2888 | 113 |
AF161554.1 | - | 4 - 5 | 632 | 113 | |
AJ238403.1 | - | 4 - 5 | 3735 | 113 | |
AY576987.1 | - | 4 - 5 | 3397 | 113 | |
AY576988.1 | - | 4 - 5 | 3397 | 113 | |
BC090954.1 | - | 4 - 5 | 2678 | 113 | |
BC117162.1 | - | 4 - 5 | 3511 | 113 | |
BC117164.1 | - | 4 - 5 | 3511 | 113 |