Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMSB10
Entrez Gene ID
9168
Gene Name
thymosin beta 10
Gene Aliases
MIG12, TB10
Chromosome Location
Chr.2: 84905639 - 84906675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021103.3 NP_066926.1 1 - 2 84 59
GenBank mRNA AK130949.1 - 1 - 2 72 59
AK311952.1 - 1 - 2 67 59
BC016025.1 - 1 - 2 49 59
BC016731.1 - 1 - 2 71 59
BC107889.1 - 1 - 2 71 59
M20259.1 - 1 - 2 49 59
S54005.1 - 1 - 2 54 59

View Full Product Details