Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMC6
Entrez Gene ID
83460
Gene Name
ER membrane protein complex subunit 6
Gene Aliases
TMEM93
Chromosome Location
Chr.17: 3668723 - 3669669 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031298.3 NP_112588.1 1 - 2 178 64
GenBank mRNA BC001409.1 - 1 - 2 86 64
BM825446.1 - 1 - 2 72 64
HY014822.1 - 1 - 2 179 64

View Full Product Details