The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032103.2 | NP_115286.1 | 1 - 2 | 351 | 58 |
NM_032104.2 | NP_115287.1 | 1 - 2 | 351 | 58 | |
XM_017001354.1 | XP_016856843.1 | 1 - 2 | 1830 | 58 | |
GenBank mRNA | AB050641.1 | - | 1 - 2 | 351 | 58 |
AB050642.1 | - | 1 - 2 | 351 | 58 | |
BX501884.1 | - | 1 - 2 | 312 | 58 |