Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC12
Entrez Gene ID
89858
Gene Name
sialic acid binding Ig like lectin 12 (gene/pseudogene)
Gene Aliases
S2V, SIGLECL1, SLG, Siglec-XII
Chromosome Location
Chr.19: 51491227 - 51501789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033329.2 NP_201586.1 1 - 2 472 81
NM_053003.3 NP_443729.1 2 - 3 862 81
XM_011527441.2 XP_011525743.1 2 - 3 559 81
XM_011527442.2 XP_011525744.1 2 - 3 862 81
GenBank mRNA AF282256.1 - 2 - 3 842 81
AK095919.1 - 2 - 3 862 81
AK225931.1 - 2 - 3 525 81
AY358140.1 - 2 - 3 820 81
BC035809.1 - 2 - 3 859 81

View Full Product Details