Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
Chromosome Location
Chr.19: 1009651 - 1021169 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033026.1 NP_001028198.1 8 - 9 1225 63
NM_033420.3 NP_219488.1 8 - 9 1225 63
XM_005259675.2 XP_005259732.1 8 - 9 1270 63
XM_005259676.2 XP_005259733.1 8 - 9 1252 63
XM_005259677.2 XP_005259734.1 8 - 9 1219 63
XM_005259678.2 XP_005259735.1 8 - 9 1270 63
XM_017027457.1 XP_016882946.1 8 - 9 1219 63
GenBank mRNA AK090400.1 - 4 - 5 290 63
AK095756.1 - 8 - 9 1225 63
BC008957.2 - 8 - 9 1198 63
DQ005958.1 - 8 - 9 1207 63

View Full Product Details