The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168303.1 | NP_001161775.1 | 1 - 2 | 357 | 108 |
GenBank mRNA | AL591986.1 | - | 1 - 2 | 154 | 108 |
DB074659.1 | - | 1 - 2 | 358 | 108 |