The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320768.1 | NP_001307697.1 | 2 - 3 | 643 | 115 |
NM_052954.3 | NP_443186.2 | 2 - 3 | 643 | 115 | |
XM_011529450.2 | XP_011527752.1 | 2 - 3 | 523 | 115 | |
GenBank mRNA | AF401639.1 | - | 2 - 3 | 248 | 115 |
AK054581.1 | - | 2 - 3 | 523 | 115 | |
AK223576.1 | - | 2 - 3 | 360 | 115 | |
AK313231.1 | - | 2 - 3 | 429 | 115 | |
AL833200.1 | - | 2 - 3 | 454 | 115 | |
AY061853.1 | - | 2 - 3 | 188 | 115 | |
BC036761.1 | - | 2 - 3 | 431 | 115 | |
JF693912.1 | - | 2 - 3 | 191 | 115 | |
JF693914.1 | - | 1 - 2 | 116 | 115 |