Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C11orf52
- Entrez Gene ID
-
91894
- Gene Name
- chromosome 11 open reading frame 52
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 111918877 - 111926871 on Build GRCh38
- Gene Symbol
- HSPB2-C11orf52
- Entrez Gene ID
-
100528019
- Gene Name
- HSPB2-C11orf52 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 111912736 - 111926871 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037651.1 |
|
2 - 3 |
511 |
96 |
GenBank mRNA |
BC110872.1 |
- |
2 - 3 |
506 |
96 |
View Full Product Details