The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004328.4 | NP_004319.1 | 2 - 3 | 292 | 136 |
GenBank mRNA | AK223518.1 | - | 1 - 2 | 169 | 136 |
BC000416.2 | - | 1 - 2 | 213 | 136 | |
BC007500.2 | - | 1 - 2 | 213 | 136 |