The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006624.1 | NP_001006625.1 | 3 - 4 | 436 | 58 |
NM_001006625.1 | NP_001006626.1 | 3 - 4 | 436 | 58 | |
NM_006474.4 | NP_006465.3 | 3 - 4 | 612 | 58 | |
NM_198389.2 | NP_938203.2 | 3 - 4 | 612 | 58 | |
XM_006710295.1 | XP_006710358.1 | 3 - 4 | 343 | 58 | |
GenBank mRNA | AB127958.1 | - | 3 - 4 | 335 | 58 |
AF030428.1 | - | 3 - 4 | 515 | 58 | |
AF390106.1 | - | 3 - 4 | 518 | 58 | |
AJ225022.1 | - | 3 - 4 | 467 | 58 | |
AK075345.1 | - | 3 - 4 | 537 | 58 | |
AK307705.1 | - | 3 - 4 | 343 | 58 | |
AK307706.1 | - | 4 - 5 | 498 | 58 | |
AK312607.1 | - | 3 - 4 | 538 | 58 | |
AY194238.1 | - | 3 - 4 | 538 | 58 | |
BC014668.1 | - | 3 - 4 | 568 | 58 | |
BC022812.1 | - | 3 - 4 | 568 | 58 | |
BQ212176.1 | - | 3 - 4 | 433 | 58 | |
BQ214601.1 | - | 3 - 4 | 365 | 58 | |
CA312590.1 | - | 3 - 4 | 58 |