Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
Chromosome Location
Chr.11: 67119210 - 67258082 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012308.2 NP_036440.1 2 - 3 908 63
NR_027473.1 2 - 3 908 63
XM_011544860.2 XP_011543162.1 2 - 3 967 63
GenBank mRNA AB023221.2 - 2 - 3 225 63
AK024505.1 - 2 - 3 150 63
BC047486.1 - 2 - 3 910 63
BC064360.1 - 2 - 3 323 63
JQ710741.1 - 1 - 2 92 63
JQ710743.1 - 1 - 2 92 63

View Full Product Details