Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP33
Entrez Gene ID
23032
Gene Name
ubiquitin specific peptidase 33
Gene Aliases
VDU1
Chromosome Location
Chr.1: 77695987 - 77759879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015017.4 NP_055832.3 4 - 5 597 74
NM_201624.2 NP_963918.1 3 - 4 418 74
NM_201626.2 NP_963920.1 4 - 5 597 74
XM_005270648.1 XP_005270705.1 3 - 4 245 74
XM_005270649.1 XP_005270706.1 3 - 4 245 74
XM_011541055.1 XP_011539357.1 3 - 4 245 74
XM_011541056.2 XP_011539358.1 3 - 4 245 74
XM_017000722.1 XP_016856211.1 3 - 4 418 74
XM_017000723.1 XP_016856212.1 3 - 4 369 74
XM_017000724.1 XP_016856213.1 3 - 4 418 74
XM_017000725.1 XP_016856214.1 3 - 4 418 74
XM_017000726.1 XP_016856215.1 3 - 4 418 74
XM_017000727.1 XP_016856216.1 3 - 4 261 74
XM_017000728.1 XP_016856217.1 3 - 4 223 74
XM_017000729.1 XP_016856218.1 3 - 4 245 74
GenBank mRNA AB029020.1 - 3 - 4 339 74
AF383172.1 - 4 - 5 570 74
AF383173.1 - 3 - 4 391 74
AK022864.1 - 4 - 5 570 74
AK308402.1 - 3 - 4 309 74
BC016663.1 - 4 - 5 451 74
DB232523.1 - 3 - 4 418 74

View Full Product Details