The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015986.3 | NP_057070.3 | 7 - 8 | 1182 | 79 |
NR_073118.1 | 6 - 7 | 974 | 79 | ||
XM_011524879.1 | XP_011523181.1 | 7 - 8 | 1356 | 79 | |
GenBank mRNA | AF046059.1 | - | 7 - 8 | 1095 | 79 |
AF120151.1 | - | 6 - 7 | 1548 | 79 | |
AK296238.1 | - | 6 - 7 | 885 | 79 | |
AK313315.1 | - | 7 - 8 | 1171 | 79 | |
BC023567.2 | - | 7 - 8 | 1104 | 79 | |
DQ298450.1 | - | 7 - 8 | 1061 | 79 |