The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016458.3 | NP_057542.2 | 5 - 6 | 1074 | 68 |
GenBank mRNA | AF151022.1 | - | 2 - 3 | 466 | 68 |
AK098226.1 | - | 6 - 7 | 768 | 68 | |
BC003035.1 | - | 5 - 6 | 1074 | 68 | |
BC009915.2 | - | 5 - 6 | 1074 | 68 | |
BC015471.1 | - | 6 - 7 | 605 | 68 |