The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193314.1 | NP_001180243.1 | 9 - 10 | 1174 | 77 |
NM_001193315.1 | NP_001180244.1 | 9 - 10 | 774 | 77 | |
NM_001193316.1 | NP_001180245.1 | 8 - 9 | 627 | 77 | |
NM_001193317.1 | NP_001180246.1 | 9 - 10 | 951 | 77 | |
NM_022067.3 | NP_071350.2 | 10 - 11 | 1045 | 77 | |
XM_017021580.1 | XP_016877069.1 | 9 - 10 | 765 | 77 | |
XM_017021581.1 | XP_016877070.1 | 9 - 10 | 899 | 77 | |
GenBank mRNA | AK022769.1 | - | 9 - 10 | 722 | 77 |
AK022925.1 | - | 9 - 10 | 700 | 77 | |
AK098254.1 | - | 9 - 10 | 1135 | 77 | |
AK222706.1 | - | 9 - 10 | 720 | 77 | |
AK297388.1 | - | 9 - 10 | 876 | 77 | |
AK298354.1 | - | 8 - 9 | 573 | 77 | |
BC015054.1 | - | 10 - 11 | 966 | 77 | |
CR457294.1 | - | 8 - 9 | 628 | 77 |