Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NADK
Entrez Gene ID
65220
Gene Name
NAD kinase
Gene Aliases
dJ283E3.1
Chromosome Location
Chr.1: 1751232 - 1780514 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198994.1 NP_001185923.1 1 - 2 166 140
XM_005244778.2 XP_005244835.1 1 - 2 244 140
XM_006710837.2 XP_006710900.1 2 - 3 359 140
XM_006710839.1 XP_006710902.1 2 - 3 359 140
XM_011542007.1 XP_011540309.1 1 - 2 244 140
XM_017002143.1 XP_016857632.1 1 - 2 244 140
GenBank mRNA AF250320.1 - 1 - 2 166 140

View Full Product Details