The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024864.4 | NP_079140.2 | 4 - 5 | 1101 | 71 |
GenBank mRNA | AK026231.1 | - | 4 - 5 | 559 | 71 |
AK225449.1 | - | 4 - 5 | 559 | 71 | |
AK292307.1 | - | 4 - 5 | 1107 | 71 | |
BC009416.1 | - | 4 - 5 | 602 | 71 | |
BC072411.1 | - | 4 - 5 | 1041 | 71 | |
CR457357.1 | - | 3 - 4 | 292 | 71 |