The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319303.1 | NP_001306232.1 | 4 - 5 | 1793 | 88 |
NM_030929.4 | NP_112191.2 | 4 - 5 | 1738 | 88 | |
NR_135067.1 | 3 - 4 | 1176 | 88 | ||
XM_017016714.1 | XP_016872203.1 | 3 - 4 | 817 | 88 | |
XM_017016715.1 | XP_016872204.1 | 3 - 4 | 817 | 88 | |
GenBank mRNA | AK172862.1 | - | 4 - 5 | 989 | 88 |
AK172864.1 | - | 4 - 5 | 1792 | 88 | |
AK225949.1 | - | 4 - 5 | 990 | 88 | |
AY359087.1 | - | 4 - 5 | 994 | 88 | |
BC007758.2 | - | 4 - 5 | 1139 | 88 | |
BI766345.1 | - | 3 - 4 | 449 | 88 | |
CX871488.1 | - | 3 - 4 | 467 | 88 |