The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033343.3 | NP_203129.1 | 1 - 2 | 306 | 128 |
GenBank mRNA | AF179849.1 | - | 1 - 2 | 310 | 128 |
AY053457.1 | - | 1 - 2 | 98 | 128 | |
BC011759.2 | - | 1 - 2 | 117 | 128 | |
BX954266.1 | - | 1 - 2 | 308 | 128 | |
KU178818.1 | - | 1 - 2 | 74 | 128 | |
KU178819.1 | - | 1 - 2 | 74 | 128 |