The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291730.1 | NP_001278659.1 | 3 - 4 | 490 | 61 |
NM_033419.4 | NP_219487.3 | 3 - 4 | 490 | 61 | |
XM_011525480.2 | XP_011523782.1 | 3 - 4 | 477 | 61 | |
GenBank mRNA | AB088396.1 | - | 3 - 4 | 445 | 61 |
AK057911.1 | - | 4 - 5 | 456 | 61 | |
AK301106.1 | - | 4 - 5 | 392 | 61 | |
AK314469.1 | - | 3 - 4 | 450 | 61 | |
AY358437.1 | - | 3 - 4 | 466 | 61 | |
BC010652.1 | - | 3 - 4 | 451 | 61 |