The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052963.2 | NP_443195.1 | 1 - 2 | 150 | 61 |
XM_005250781.3 | XP_005250838.1 | 1 - 2 | 138 | 61 | |
GenBank mRNA | AF349017.1 | - | 1 - 2 | 139 | 61 |
AK303237.1 | - | 1 - 2 | 111 | 61 | |
AK308475.1 | - | 1 - 2 | 122 | 61 | |
AK311647.1 | - | 1 - 2 | 122 | 61 | |
AK315913.1 | - | 1 - 2 | 112 | 61 | |
BC044646.1 | - | 1 - 2 | 125 | 61 | |
BC052285.1 | - | 1 - 2 | 125 | 61 | |
BC071914.1 | - | 1 - 2 | 123 | 61 | |
CB149603.1 | - | 1 - 2 | 140 | 61 | |
DC375680.1 | - | 1 - 2 | 150 | 61 |