The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080430.2 | NP_536355.1 | 2 - 3 | 230 | 92 |
GenBank mRNA | AY043487.1 | - | 2 - 3 | 259 | 92 |
BC013421.1 | - | 2 - 3 | 176 | 92 | |
BC030236.1 | - | 2 - 3 | 187 | 92 | |
BC068004.1 | - | 2 - 3 | 220 | 92 |