Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRD
Entrez Gene ID
5789
Gene Name
protein tyrosine phosphatase, receptor type D
Gene Aliases
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location
Chr.9: 8314246 - 10612840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040712.2 NP_001035802.1 1 - 2 164 67
NM_001171025.1 NP_001164496.1 1 - 2 164 67
NM_002839.3 NP_002830.1 12 - 13 804 67
NM_130391.3 NP_569075.2 1 - 2 164 67
NM_130392.3 NP_569076.2 1 - 2 164 67
NM_130393.3 NP_569077.2 1 - 2 164 67
XM_006716817.3 XP_006716880.1 11 - 12 1102 67
XM_006716823.2 XP_006716886.1 11 - 12 1096 67
XM_006716825.3 XP_006716888.1 11 - 12 1098 67
XM_006716827.3 XP_006716890.1 11 - 12 1095 67
XM_006716832.3 XP_006716895.1 11 - 12 1090 67
XM_006716833.3 XP_006716896.1 11 - 12 1092 67
XM_006716834.3 XP_006716897.1 11 - 12 1093 67
XM_006716835.3 XP_006716898.1 11 - 12 1092 67
XM_006716837.3 XP_006716900.1 11 - 12 1090 67
XM_006716838.3 XP_006716901.1 11 - 12 1092 67
XM_006716839.3 XP_006716902.1 11 - 12 1090 67
XM_011517992.2 XP_011516294.1 11 - 12 1095 67
XM_017014958.1 XP_016870447.1 11 - 12 1101 67
XM_017014959.1 XP_016870448.1 12 - 13 1173 67
XM_017014960.1 XP_016870449.1 10 - 11 1062 67
XM_017014961.1 XP_016870450.1 10 - 11 1046 67
XM_017014962.1 XP_016870451.1 2 - 3 234 67
XM_017014963.1 XP_016870452.1 11 - 12 1101 67
XM_017014964.1 XP_016870453.1 11 - 12 1100 67
XM_017014965.1 XP_016870454.1 11 - 12 1103 67
XM_017014966.1 XP_016870455.1 11 - 12 1101 67
XM_017014967.1 XP_016870456.1 11 - 12 1099 67
XM_017014968.1 XP_016870457.1 11 - 12 1098 67
XM_017014969.1 XP_016870458.1 11 - 12 1101 67
XM_017014970.1 XP_016870459.1 11 - 12 1101 67
XM_017014971.1 XP_016870460.1 11 - 12 1099 67
XM_017014972.1 XP_016870461.1 11 - 12 1101 67
XM_017014973.1 XP_016870462.1 11 - 12 1098 67
XM_017014974.1 XP_016870463.1 11 - 12 1101 67
XM_017014975.1 XP_016870464.1 11 - 12 1096 67
XM_017014976.1 XP_016870465.1 11 - 12 1098 67
XM_017014977.1 XP_016870466.1 11 - 12 1099 67
XM_017014978.1 XP_016870467.1 11 - 12 1098 67
XM_017014979.1 XP_016870468.1 11 - 12 1100 67
XM_017014980.1 XP_016870469.1 11 - 12 1095 67
XM_017014981.1 XP_016870470.1 11 - 12 1090 67
XM_017014982.1 XP_016870471.1 11 - 12 1097 67
XM_017014983.1 XP_016870472.1 11 - 12 1097 67
XM_017014984.1 XP_016870473.1 11 - 12 1097 67
XM_017014985.1 XP_016870474.1 11 - 12 1094 67
XM_017014986.1 XP_016870475.1 11 - 12 1093 67
XM_017014987.1 XP_016870476.1 11 - 12 1093 67
XM_017014988.1 XP_016870477.1 11 - 12 1093 67
XM_017014989.1 XP_016870478.1 11 - 12 1093 67
XM_017014990.1 XP_016870479.1 11 - 12 1092 67
XM_017014991.1 XP_016870480.1 11 - 12 1092 67
XM_017014992.1 XP_016870481.1 11 - 12 1092 67
XM_017014993.1 XP_016870482.1 11 - 12 1090 67
XM_017014994.1 XP_016870483.1 11 - 12 1090 67
XM_017014995.1 XP_016870484.1 11 - 12 1096 67
GenBank mRNA AB211400.1 - 11 - 12 772 67
BC028038.1 - - 876 67
BC106713.1 - 1 - 2 112 67
BC106714.1 - 1 - 2 112 67
BC106715.2 - 1 - 2 112 67
BC106716.1 - 1 - 2 112 67
DR001889.1 - 3 - 4 239 67
L38929.1 - 2 - 3 214 67

View Full Product Details