The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143688.2 | NP_001137160.1 | 9 - 10 | 1346 | 131 |
NM_001323936.1 | NP_001310865.1 | 9 - 10 | 1265 | 131 | |
NM_001323937.1 | NP_001310866.1 | 8 - 9 | 1167 | 131 | |
NM_001323938.1 | NP_001310867.1 | 9 - 10 | 1273 | 131 | |
NM_001323939.1 | NP_001310868.1 | 10 - 11 | 1470 | 131 | |
NM_001323940.1 | NP_001310869.1 | 8 - 9 | 1176 | 131 | |
NM_001323941.1 | NP_001310870.1 | 9 - 10 | 1741 | 131 | |
NM_001323943.1 | NP_001310872.1 | 8 - 9 | 1370 | 131 | |
NM_001323944.1 | NP_001310873.1 | 9 - 10 | 1540 | 131 | |
NM_001323945.1 | NP_001310874.1 | 9 - 10 | 1814 | 131 | |
NM_001323946.1 | NP_001310875.1 | 9 - 10 | 1467 | 131 | |
NM_001323948.1 | NP_001310877.1 | 8 - 9 | 1411 | 131 | |
NM_133375.4 | NP_588616.1 | 9 - 10 | 1296 | 131 | |
XM_005254146.3 | XP_005254203.1 | 9 - 10 | 1414 | 131 | |
GenBank mRNA | AB075835.1 | - | 8 - 9 | 1003 | 131 |
AK095407.1 | - | 9 - 10 | 1325 | 131 | |
AK295392.1 | - | 8 - 9 | 1003 | 131 | |
BC014124.2 | - | 1 - 2 | 94 | 131 | |
BC022089.2 | - | 9 - 10 | 1231 | 131 |