The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135774.1 | NP_001129246.1 | 3 - 4 | 633 | 71 |
NM_003490.3 | NP_003481.3 | 2 - 3 | 530 | 71 | |
NM_133633.2 | NP_598344.2 | 2 - 3 | 530 | 71 | |
XM_011530405.2 | XP_011528707.1 | 3 - 4 | 1166 | 71 | |
XM_011530406.2 | XP_011528708.1 | 3 - 4 | 632 | 71 | |
XM_011530407.2 | XP_011528709.1 | 3 - 4 | 1039 | 71 | |
XM_011530408.1 | XP_011528710.1 | 3 - 4 | 630 | 71 | |
XM_017028960.1 | XP_016884449.1 | 4 - 5 | 1564 | 71 | |
XM_017028961.1 | XP_016884450.1 | 3 - 4 | 956 | 71 | |
XM_017028962.1 | XP_016884451.1 | 3 - 4 | 891 | 71 | |
XM_017028963.1 | XP_016884452.1 | 3 - 4 | 555 | 71 | |
XM_017028964.1 | XP_016884453.1 | 3 - 4 | 630 | 71 | |
GenBank mRNA | AB209684.1 | - | 3 - 4 | 585 | 71 |
AF046873.1 | - | 2 - 3 | 407 | 71 | |
BC075065.2 | - | 2 - 3 | 397 | 71 | |
BC075066.2 | - | 2 - 3 | 397 | 71 | |
BC117460.1 | - | 2 - 3 | 398 | 71 | |
BC143874.1 | - | 2 - 3 | 398 | 71 | |
CR456589.1 | - | 2 - 3 | 462 | 71 |